5-Iodo-1-methylimidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Iodo-1-methyl-1H-imidazole
OR10512-1G
OR10512-5G
APOSOR10512-1GEA
74.1
EUR
APOSOR10512-1G
APOSOR10512-5G
5-Iodo-1-methylimidazole
5-Iodo-1-methylimidazole
Formel:
C₄H₅IN₂ Molecular Weight: 208,00 g/mol Schmelzpunkt: 105…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632215 CAS-Nummer: 71759-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...