4-Iodimidazol 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Iod-1H-imidazol

OR10517-25G OR10517-5G OR10517-100G
APOSOR10517-25GEA 72.5 EUR
APOSOR10517-25G APOSOR10517-5G APOSOR10517-100G
4-Iodimidazol 96%
4-Iodoimidazole
Formel: C₃H₃IN₂
Molecular Weight: 193,97 g/mol
Schmelzpunkt: 135…138 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01632213
CAS-Nummer: 71759-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR