2-Isopropylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Isopropylbenzeneboronic acid, (2-Isopropylphenyl)boronic acid

OR10522-1G OR10522-5G OR10522-25G
APOSOR10522-1GEA 21.9 EUR
APOSOR10522-1G APOSOR10522-5G APOSOR10522-25G
2-Isopropylphenylboronsäure
2-Isopropylphenylboronsäure
Formel: C₉H₁₃BO₂
Molecular Weight: 164,01 g/mol
Schmelzpunkt: 74…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411937
CAS-Nummer: 89787-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR