3-(Methoxycarbonyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-boronobenzoate, 3-(Methoxycarbonyl)benzeneboronic acid

OR10536-5G OR10536-1G
APOSOR10536-5GEA 24.8 EUR
APOSOR10536-5G APOSOR10536-1G
3-(Methoxycarbonyl)phenylboronsäure 97%
3-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179,97 g/mol
Schmelzpunkt: 205…208 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02093046
CAS-Nummer: 99769-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR