Methyl-7-indolcarboxylat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1H-indole-7-carboxylate

OR10545-25G OR10545-5G
APOSOR10545-25GEA 98.6 EUR
APOSOR10545-25G APOSOR10545-5G
Methyl-7-indolcarboxylat 97%
Methyl-7-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175.19 g/mol
Schmelzpunkt: 47…48 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00211064
CAS-Nummer: 93247-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR