4-(Methylcarbamoyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methylcarbamoyl)benzeneboronic acid, 4-(N-Methylaminocarbonyl)phenylboronic acid

OR10555-1G OR10555-5G
APOSOR10555-1GEA 21.9 EUR
APOSOR10555-1G APOSOR10555-5G
4-(Methylcarbamoyl)phenylboronsäure
4-(Methylcarbamoyl)phenylboronsäure
Formel: C₈H₁₀BNO₃
Molecular Weight: 178.98 g/mol
Schmelzpunkt: 200…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788427
CAS-Nummer: 121177-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR