2-(Methylsulphonylamino)benzeneboronic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-(Methylsulphonamido)phenyl)boronic acid
OR10560-1G
OR10560-5G
APOSOR10560-1GEA
91.5
EUR
APOSOR10560-1G
APOSOR10560-5G
2-(Methylsulphonylamino)benzeneboronic acid 97%
2-(Methylsulphonylamino)benzeneboronic acid
Formel:
C₇H₁₀BNO₄S Molecular Weight: 215.03 g/mol Schmelzpunkt: 116…120 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179471 CAS-Nummer: 756520-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...