3-(Methylsulphonylamino)phenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methylsulphonylaminophenylboronic acid, 3-(Methylsulphonylamino)phenylboronic Acid

OR10561-1G
APOSOR10561-1GEA 21.9 EUR
APOSOR10561-1G
3-(Methylsulphonylamino)phenylboronic acid 98%
3-(Methylsulphonylamino)phenylboronic acid
Formel: C₇H₁₀BNO₄S
Molecular Weight: 215,04 g/mol
Schmelzpunkt: 90…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179478
CAS-Nummer: 148355-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR