4-(4-Morpholinylcarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid

OR10568-1G OR10568-5G OR10568-25G
APOSOR10568-1GEA 120 EUR
APOSOR10568-1G APOSOR10568-5G APOSOR10568-25G
4-(4-Morpholinylcarbonyl)phenylboronsäure 98%
4-(4-Morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₄
Molecular Weight: 235,05 g/mol
Schmelzpunkt: 126…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411952
CAS-Nummer: 389621-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR