4-(Piperidin-1-carbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(Piperidin-1-yl)carbonyl]benzeneboronic acid

OR10582-1G OR10582-100MG
APOSOR10582-1GEA 79.9 EUR
APOSOR10582-1G APOSOR10582-100MG
4-(Piperidin-1-carbonyl)phenylboronsäure
4-(Piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₆BNO₃
Schmelzpunkt: 150…154 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411951
CAS-Nummer: 389621-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR