Tetrahydropyran-4-carbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tetrahydropyran-4-yl-carboxylic acid
OR10587-25G
OR10587-5G
OR10587-100G
APOSOR10587-25GEA
31.9
EUR
APOSOR10587-25G
APOSOR10587-5G
APOSOR10587-100G
Tetrahydropyran-4-carbonsäure 98%
Tetrahydropyran-4-carbonsäure
Formel:
C₆H₁₀O₃ Molecular Weight: 130,14 g/mol Schmelzpunkt: 87…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031016 CAS-Nummer: 5337-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...