ɑ-[2-(Methylamino)ethyl]benzyl alcohol 98%
Lieferant: Apollo Scientific
Synonyme:
3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol, α-[2-(Methylamino)ethyl]benzyl Alcohol, 3-(Methylamino)-1-phenylpropan-1-ol
OR1059-500G
APOSOR1059-500GEA
168
EUR
APOSOR1059-500G
ɑ-[2-(Methylamino)ethyl]benzyl alcohol 98%
alpha-[2-(Methylamino)ethyl]benzyl alcohol
Formel:
C₁₀H₁₅NO Molecular Weight: 165,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674078 CAS-Nummer: 42142-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...