1-Benzyl-1H-pyrazol-4-boronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Phenylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR10602-1G
OR10602-25G
OR10602-5G
APOSOR10602-1GEA
21.9
EUR
APOSOR10602-1G
APOSOR10602-25G
APOSOR10602-5G
1-Benzyl-1H-pyrazol-4-boronsäurepinakolester
1-Benzyl-1H-pyrazol-4-boronsäurepinakolester
Formel:
C₁₆H₂₁BN₂O₂ Molecular Weight: 284,16 g/mol Schmelzpunkt: 86…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03789252 CAS-Nummer: 761446-45-1 EINECS: 261-911-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...