3-Phenylbenzoesäure 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

OR10604-1G OR10604-25G OR10604-5G
APOSOR10604-1GEA 21.9 EUR
APOSOR10604-1G APOSOR10604-25G APOSOR10604-5G
3-Phenylbenzoesäure 98+%
3-Phenylbenzoic acid
Formel: C₁₃H₁₀O₂
Molecular Weight: 198.22 g/mol
Schmelzpunkt: 164…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045846
CAS-Nummer: 716-76-7
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR