Ethyl-(triphenylphosphoranyliden)acetat 94%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate

OR10645-100G OR10645-25G OR10645-500G
APOSOR10645-100GEA 29 EUR
APOSOR10645-100G APOSOR10645-25G APOSOR10645-500G
Ethyl-(triphenylphosphoranyliden)acetat 94%
Ethyl-2-(triphenylphosphoranylidene)acetate
Formel: C₂₂H₂₁O₂P
Molecular Weight: 348,38 g/mol
Schmelzpunkt: 128…130 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009183
CAS-Nummer: 1099-45-2
EINECS: 214-151-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR