(Triphenylphosphoranyliden)acetaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Triphenylphosphoranylidene)acetaldehyde, (Formylmethylene)triphenylphosphorane
OR10653-25G
OR10653-100G
APOSOR10653-25GEA
21.9
EUR
APOSOR10653-25G
APOSOR10653-100G
(Triphenylphosphoranyliden)acetaldehyd 95%
(Triphenylphosphoranylidene)acetaldehyde
Formel:
C₂₀H₁₇OP Molecular Weight: 304.33 g/mol Schmelzpunkt: 185…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006994 CAS-Nummer: 2136-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...