3-tert-Butyl-1-brombenzol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3-Bromphenyl)-2-methylpropan, 3-Brom-tert-butylbenzol, 1-Brom-3-tert-butylbenzol
OR1066-100G
OR1066-25G
OR1066-5G
APOSOR1066-100GEA
304
EUR
APOSOR1066-100G
APOSOR1066-25G
APOSOR1066-5G
3-tert-Butyl-1-brombenzol 99%
3-tert-Butyl-1-brombenzol
Formel:
C₁₀H₁₃Br Molecular Weight: 213,12 g/mol Siedepunkt: 95…98 °C/10mmHg Dichte: 1,251 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234661 CAS-Nummer: 3972-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...