3-Brom-4-hydroxybenzonitril

Lieferant: Apollo Scientific
Achtung

OR1074-100G OR1074-25G OR1074-5G
APOSOR1074-100GEA 128 EUR
APOSOR1074-100G APOSOR1074-25G APOSOR1074-5G
3-Brom-4-hydroxybenzonitril
3-Brom-4-hydroxybenzonitril
Formel: C₇H₄BrNO
Molecular Weight: 198,02 g/mol
Schmelzpunkt: 155…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143096
CAS-Nummer: 2315-86-8
EINECS: 219-022-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR