Dimethyl-2-oxo-4-phenylbutylphosphonate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dimethyl (2-oxo-4-phenylbutyl)phosphonate
OR10775-1G
OR10775-5G
APOSOR10775-1GEA
91.5
EUR
APOSOR10775-1G
APOSOR10775-5G
Dimethyl-2-oxo-4-phenylbutylphosphonate
Dimethyl-2-oxo-4-phenylbutylphosphonate
Formel:
C₁₂H₁₇O₄P Molecular Weight: 256.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093040 CAS-Nummer: 41162-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...