Tetraethyl-p-xylylenediphosphonate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
p-Xylylenediphosphonic Acid Tetraethyl Ester, p-Bis(diethylphosphono)xylene
OR10800-5G
OR10800-100G
OR10800-25G
APOSOR10800-5GEA
108
EUR
APOSOR10800-5G
APOSOR10800-100G
APOSOR10800-25G
Tetraethyl-p-xylylenediphosphonate 98%
Tetraethyl-p-xylylenediphosphonate
Formel:
C₁₆H₂₈O₆P₂ Molecular Weight: 378,34 g/mol Siedepunkt: 220 °C/7mmHg Schmelzpunkt: 78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671576 CAS-Nummer: 4546-04-7 EINECS: 224-902-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...