(4-Aminophenyl)essigsäureethylester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl-(4-aminophenyl)acetat

OR10871-100G OR10871-25G
APOSOR10871-100GEA 157 EUR
APOSOR10871-100G APOSOR10871-25G
(4-Aminophenyl)essigsäureethylester 98%
(4-Aminophenyl)essigsäureethylester
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017569
CAS-Nummer: 5438-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR