Monoethyl itaconate
Lieferant: Apollo Scientific
Synonyme:
beta-Ethyl Itaconate, 4-Ethoxy-2-methylene-4-oxobutanoic acid, Itaconic Acid Monoethyl Ester, 4-Ethyl Methylenesuccinate, Ethyl itaconate
OR10872-25G
OR10872-100G
APOSOR10872-25GEA
105
EUR
APOSOR10872-25G
APOSOR10872-100G
Monoethyl itaconate
Monoethyl itaconate
Formel:
C₇H₁₀O₄ Molecular Weight: 158,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059138 CAS-Nummer: 57718-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...