2-(4-Methanesulphonamidophenyl)acetic acid 97+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Methylsulphonylamino)phenylacetic Acid
OR10894-1G
OR10894-5G
APOSOR10894-1GEA
174
EUR
APOSOR10894-1G
APOSOR10894-5G
2-(4-Methanesulphonamidophenyl)acetic acid 97+%
2-(4-Methanesulphonamidophenyl)acetic acid
Formel:
C₉H₁₁NO₄S Molecular Weight: 229.26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411195 CAS-Nummer: 56205-88-0 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...