3,4-Dichlorbenzonitril 98%

Lieferant: Apollo Scientific
Gefahr

OR10910-100G OR10910-500G
APOSOR10910-100GEA 85.7 EUR
APOSOR10910-100G APOSOR10910-500G
3,4-Dichlorbenzonitril 98%
3,4-Dichlorbenzonitril
Formel: C₇H₃Cl₂N
Molecular Weight: 172,01 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016379
CAS-Nummer: 6574-99-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR