3,5-Dimethylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid

OR10931-25G OR10931-5G OR10931-100G
APOSOR10931-25GEA 37.7 EUR
APOSOR10931-25G APOSOR10931-5G APOSOR10931-100G
3,5-Dimethylphenylboronsäure
3,5-Dimethylphenylboronsäure
Formel: C₈H₁₁BO₂
Molecular Weight: 149,98 g/mol
Schmelzpunkt: 235…237 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00185689
CAS-Nummer: 172975-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR