5-Chlor-2-indolcarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-1H-indole-2-carboxylic acid
OR10933-25G
APOSOR10933-25GEA
98.6
EUR
APOSOR10933-25G
5-Chlor-2-indolcarbonsäure
5-Chloroindole-2-carboxylic acid
Formel:
C₉H₆ClNO₂ Molecular Weight: 195,6 g/mol Schmelzpunkt: 287 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005613 CAS-Nummer: 10517-21-2 EINECS: 234-050-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...