(4-Aminophenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Aminophenylacetic acid

OR10955-500G
APOSOR10955-500GEA 231 EUR
APOSOR10955-500G
(4-Aminophenyl)essigsäure
(4-Aminophenyl)essigsäure
Formel: (C₆H₄(NH₂))CH₂COOH
Molecular Weight: 151,16 g/mol
Schmelzpunkt: 201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007916
CAS-Nummer: 1197-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR