2-Amino-4,6-dimethoxypyrimidin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,6-Dimethoxypyrimidin-2-amine, 4,6-Dimethoxypyrimidin-2-ylamine
OR10960-100G
OR10960-25G
APOSOR10960-100GEA
119
EUR
APOSOR10960-100G
APOSOR10960-25G
2-Amino-4,6-dimethoxypyrimidin
2-Amino-4,6-dimethoxypyrimidine
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,16 g/mol Schmelzpunkt: 94…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038832 CAS-Nummer: 36315-01-2 EINECS: 252-969-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...