3-Amino-3-(4-hydroxyphenyl)propionsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-3-(4-hydroxyphenyl)propanoic Acid

OR110036-1G OR110036-5G OR110036-25G
APOSOR110036-1GEA 21.9 EUR
APOSOR110036-1G APOSOR110036-5G APOSOR110036-25G
3-Amino-3-(4-hydroxyphenyl)propionsäure
3-Amino-3-(4-hydroxyphenyl)propionsäure
Formel: C₉H₁₁NO₃
Molecular Weight: 181.1885 g/mol
Schmelzpunkt: 193…195 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00181810
CAS-Nummer: 6049-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR