6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid

Lieferant: Apollo Scientific

OR110118-1G OR110118-250MG
APOSOR110118-1GEA 219 EUR
APOSOR110118-1G APOSOR110118-250MG
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₈H₅N₂ClO₂
Molecular Weight: 196,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07021509
CAS-Nummer: 138642-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR