(4-Amino-3-bromophenyl)acetonitrile
Lieferant: Apollo Scientific
Synonyme:
2-(4-Amino-3-bromophenyl)acetonitrile
OR110126-1G
OR110126-250MG
OR110126-500MG
OR110126-2.5G
OR110126-5G
APOSOR110126-1GEA
102
EUR
APOSOR110126-1G
APOSOR110126-250MG
APOSOR110126-500MG
APOSOR110126-2.5G
APOSOR110126-5G
(4-Amino-3-bromophenyl)acetonitrile
(4-Amino-3-bromophenyl)acetonitrile
Formel:
C₈H₇BrN₂ Molecular Weight: 211,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275180 CAS-Nummer: 882855-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...