4-(1-Pyrrolyl)benzonitril
Lieferant: Apollo Scientific
OR110146-10G
OR110146-1G
OR110146-5G
APOSOR110146-10GEA
0
EUR
APOSOR110146-10G
APOSOR110146-1G
APOSOR110146-5G
4-(1-Pyrrolyl)benzonitril
4-(1-Pyrrolyl)benzonitril
Formel:
C₁₁H₈N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085164 CAS-Nummer: 23351-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...