Methyl-2-amino-5-methylbenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-amino-5-methylbenzoate
OR110165-5G
OR110165-25G
APOSOR110165-5GEA
21.9
EUR
APOSOR110165-5G
APOSOR110165-25G
Methyl-2-amino-5-methylbenzoat
Methyl-2-amino-5-methylbenzoat
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,1900 g/mol Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234606 CAS-Nummer: 18595-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...