1,3-Benzoxazol-5-ylacetic acid

Lieferant: Apollo Scientific
Achtung

OR110170-1G OR110170-500MG
APOSOR110170-1GEA 226 EUR
APOSOR110170-1G APOSOR110170-500MG
1,3-Benzoxazol-5-ylacetic acid
1,3-Benzoxazol-5-ylacetic acid
Formel: C₉H₇NO₃
Molecular Weight: 177.16 g/mol
Schmelzpunkt: 161…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04125031
CAS-Nummer: 153810-37-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR