Methyl-1,3-benzoxazole-7-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl benzo[d]oxazole-7-carboxylate
OR110172-1G
OR110172-250MG
APOSOR110172-1GEA
71.2
EUR
APOSOR110172-1G
APOSOR110172-250MG
Methyl-1,3-benzoxazole-7-carboxylate
Methyl-1,3-benzoxazole-7-carboxylate
Formel:
C₉H₇NO₃ Molecular Weight: 177.16 g/mol Schmelzpunkt: 114…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11042246 CAS-Nummer: 1086378-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...