6-Amino-3-chlor-1H-indazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-1H-indazol-6-amine

OR110175-1G OR110175-5G
APOSOR110175-1GEA 58 EUR
APOSOR110175-1G APOSOR110175-5G
6-Amino-3-chlor-1H-indazol
6-Amino-3-chloro-1H-indazole
Formel: C₇H₆ClN₃
Molecular Weight: 167,60 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07781648
CAS-Nummer: 21413-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR