Ethyl-4-chloro-1-benzothiophene-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR110189-10G
OR110189-1G
OR110189-5G
APOSOR110189-10GEA
335
EUR
APOSOR110189-10G
APOSOR110189-1G
APOSOR110189-5G
Ethyl-4-chloro-1-benzothiophene-2-carboxylate
Ethyl-4-chloro-1-benzothiophene-2-carboxylate
Formel:
C₁₁H₉ClO₂S Molecular Weight: 240,71 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100113 CAS-Nummer: 1207537-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...