Ethyl-3-amino-1-benzothiophene-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 3-aminobenzo[b]thiophene-2-carboxylate
OR110192-5G
OR110192-25G
APOSOR110192-5GEA
88.6
EUR
APOSOR110192-5G
APOSOR110192-25G
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Formel:
C₁₁H₁₁NO₂S Molecular Weight: 221,28 g/mol Schmelzpunkt: 82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00661903 CAS-Nummer: 34761-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...