1-(4-Chlorphenyl)biguanid Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR1102-1G OR1102-5G
APOSOR1102-1GEA 21.9 EUR
APOSOR1102-1G APOSOR1102-5G
1-(4-Chlorphenyl)biguanid Hydrochlorid
1-(4-Chlorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₁Cl₂N₅
Molecular Weight: 248.12 g/mol
Schmelzpunkt: 247…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053020
CAS-Nummer: 4022-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR