3-(2,4-Dimethylphenyl)-1H-pyrazole

Lieferant: Apollo Scientific

OR110210-1G OR110210-5G
APOSOR110210-1GEA 92.8 EUR
APOSOR110210-1G APOSOR110210-5G
3-(2,4-Dimethylphenyl)-1H-pyrazole
3-(2,4-Dimethylphenyl)-1H-pyrazole
Formel: C₁₁H₁₂N₂
Molecular Weight: 172.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505059
CAS-Nummer: 154258-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR