Imidazo[1,2-a]pyrimidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR110222-1G OR110222-250MG OR110222-500MG OR110222-2.5G
APOSOR110222-1GEA 236 EUR
APOSOR110222-1G APOSOR110222-250MG APOSOR110222-500MG APOSOR110222-2.5G
Imidazo[1,2-a]pyrimidine-3-carboxylic acid
Imidazo[1,2-a]pyrimidine-3-carboxylic acid
Formel: C₇H₅N₃O₂
Molecular Weight: 163.14 g/mol
Schmelzpunkt: 231…233 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD09994697
CAS-Nummer: 64951-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR