4-Pyridinboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR11025-25G OR11025-100G
APOSOR11025-25GEA 102 EUR
APOSOR11025-25G APOSOR11025-100G
4-Pyridinboronsäurepinakolester
4-Pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₂
Molecular Weight: 205.06 g/mol
Schmelzpunkt: 153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319051
CAS-Nummer: 181219-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR