1,3-Benzothiazole-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110285-10G OR110285-1G OR110285-5G
APOSOR110285-10GEA 377 EUR
APOSOR110285-10G APOSOR110285-1G APOSOR110285-5G
1,3-Benzothiazole-2-carbohydrazide
1,3-Benzothiazole-2-carbohydrazide
Formel: C₈H₇N₃OS
Molecular Weight: 193.23 g/mol
Schmelzpunkt: 179 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02713577
CAS-Nummer: 28891-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR