2-Hydrazino-5-nitropyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydrazinyl-5-nitropyridine
OR110327-1G
OR110327-5G
APOSOR110327-1GEA
34.8
EUR
APOSOR110327-1G
APOSOR110327-5G
2-Hydrazino-5-nitropyridine
2-Hydrazino-5-nitropyridine
Formel:
C₅H₆N₄O₂ Molecular Weight: 154,13 g/mol Schmelzpunkt: 209…210 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00118332 CAS-Nummer: 6343-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...