Kalium-tert-butoxid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Kalium-tert-butanolat, 2-Methyl-2-propanol Kaliumsalz, Kalium-tert-butylat
OR11033-100G
OR11033-500G
OR11033-1KG
APOSOR11033-100GEA
102
EUR
APOSOR11033-100G
APOSOR11033-500G
APOSOR11033-1KG
Kalium-tert-butoxid 97%
Kalium-tert-butanolat
Formel:
C₄H₉KO Molecular Weight: 112.21 g/mol Schmelzpunkt: 255 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012162 CAS-Nummer: 865-47-4 EINECS: 212-740-3 UN: 3206 ADR: 4.2+8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...