5-Bromo-3-chloropyrazin-2-amine

Lieferant: Apollo Scientific

OR110341-25G
APOSOR110341-25GEA 63.8 EUR
APOSOR110341-25G
5-Bromo-3-chloropyrazin-2-amine
5-Bromo-3-chloropyrazin-2-amine
Formel: C₄H₃BrClN₃
Molecular Weight: 208,4500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109828
CAS-Nummer: 21943-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR