5-Methoxy-1H-indazol

Lieferant: Apollo Scientific

OR110371-1G OR110371-250MG
APOSOR110371-1GEA 120 EUR
APOSOR110371-1G APOSOR110371-250MG
5-Methoxy-1H-indazol
5-Methoxy-1H-indazole
Formel: C₈H₈N₂O
Molecular Weight: 148,1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781657
CAS-Nummer: 94444-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR