6-Chloro-2-Methyl-4-Pyrimidinol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Chloro-4-hydroxy-2-methylpyrimidine, 6-Chloro-2-methylpyrimidin-4(1H)-one

OR110379-1G OR110379-5G
APOSOR110379-1GEA 139 EUR
APOSOR110379-1G APOSOR110379-5G
6-Chloro-2-Methyl-4-Pyrimidinol
6-Chloro-2-methyl-4-pyrimidinol
Formel: C₅H₅ClN₂O
Molecular Weight: 144.558 g/mol
Schmelzpunkt: 206…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00030773
CAS-Nummer: 17551-52-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR