4-Iod-3,5-xylenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Hydroxy-2-iod-3-xylol, 4-Iod-3,5-dimethylphenol, 3,5-Dimethyl-4-iodphenol

OR1104-1G OR1104-5G OR1104-25G
APOSOR1104-1GEA 26.1 EUR
APOSOR1104-1G APOSOR1104-5G APOSOR1104-25G
4-Iod-3,5-xylenol
4-Iod-3,5-xylenol
Formel: C₈H₉IO
Molecular Weight: 248.06 g/mol
Schmelzpunkt: 129…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00174284
CAS-Nummer: 80826-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR