Methyl-4-(hydroxymethyl)benzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-(hydroxymethyl)benzoate

OR110419-10G OR110419-5G OR110419-25G OR110419-100G
APOSOR110419-10GEA 24.8 EUR
APOSOR110419-10G APOSOR110419-5G APOSOR110419-25G APOSOR110419-100G
Methyl-4-(hydroxymethyl)benzoat
Methyl-4-(hydroxymethyl)benzoat
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 46…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009984
CAS-Nummer: 6908-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR